missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | Live Cell Detection, Cell Labeling |
| For Use With (Equipment) | Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells |
| Quantity | 100 mg |
For research use only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?