missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks™ Hinged Rack Barrier Tips

Product Code. 10794182 Shop All Corning Products
Click to view available options
Volume (Metric):
100 to 1000 μL
Unit Size:
Pack of 3072
This item is not returnable. View return policy

Product Code. 10794182

Brand: Corning™ 4140

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures.

 

  • The hinged rack design allows easy rack stacking
  • Racks can be reused
  • Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Used for quantitative assays in which sample binding can significantly alter results
  • These sterile tips will not trap liquids or inhibit PCR in the event of over-pipetting

 

 

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 1000 μL
Filtered Filtered
Sterility Sterile
Color Natural
Clean Claims Nonpyrogenic, Human DNA-free, RNase-/DNase-free, Sterility Assurance Level 10-3
For Use With (Application) Used for quantitative assays in which sample binding can significantly alter results
Format Hinged Rack
Tip Style Round
Packaging 96 Tips/Rack, 8 Racks/Pack
Graduated Yes
Product Line DeckWorks
Material HDPE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.