missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-yl)propanoic acid, ≥97%, Thermo Scientific™

Product Code. 10390503
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10390503

Brand: Thermo Scientific Maybridge MO01250CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 18999-45-6
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD02731119
InChI Key VSFNAZLYGOOSEY-UHFFFAOYSA-N
Synonym 3-1h-imidazol-1-yl propanoic acid, 3-imidazol-1-yl-propionic acid, 1h-imidazole-1-propanoic acid, 3-1-imidazolyl propanoic acid, chembl20724, 3-imidazolylpropanoic acid, 3-imidazol-1-yl-propionicacid, 3-1-imidazolyl propionic acid, 3 1h-imidazol-1-yl propanoic acid
PubChem CID 2794718
IUPAC Name 3-imidazol-1-ylpropanoic acid
SMILES C1=CN(C=N1)CCC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 3-(1H-imidazol-1-yl)propanoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.