Learn More
Corning™ Costar™ Tip Loading System Refills
Product Code. 11793994 Shop All Corning Products
Click to view available options
Color:
Natural
Yellow
Unit Size:
Pack of 4800
Product Code. 11793994
Brand: Corning™ 4783
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
System consists of a one-piece base which attaches to a 'magazine' consisting of 10 layers of 96 tips
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Color Natural
Format Rack
Product Line Costar