missing translation for 'onlineSavingsMsg'
Learn More

Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
BIOPHARMA

Product Code. 15363651
Change view
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
20 L
6 x 1 L
1 x 100 L
1 x 200 L
Packaging:
Bioprocess Container
Bottle
Unit Size:
100 L
20 L
200 L
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15363651 6 x 500 mL Bottle Pack of 6
15353651 6 x 100 mL Bottle Pack of 6
15373581 6 x 1 L Bottle Pack of 6
15383651 20 L Bioprocess Container 20 L
15393651 1 x 100 L Bioprocess Container 100 L
15303661 1 x 200 L Bottle 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Product Code. 15363651 Supplier Corning Media Tech Supplier No. 25055CV
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.

Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).

Order Info

Supplied in a plastic bottle except MT-25-055-LB and -LH, which come in flexible media bags.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottle
Quantity 6 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.