missing translation for 'onlineSavingsMsg'
Learn More

Tris Base Buffer, Powder, Corning™

Product Code. 15383761
Click to view available options
Quantity:
1 kg
Unit Size:
1000g
This item is not returnable. View return policy

Product Code. 15383761

Brand: Corning Media Tech 61233RR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in buffer solutions in biochemical and molecular biological research

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • pH: 7.0 to 9.2
  • Molecular weight: 121.14

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Base Buffer
pH 7 to 9.2
Physical Form Powder
Quantity 1 kg
Packaging Plastic Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.