missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Sodium Bicarbonate Solution

Description
Technical Advantage: Product manufactured under industry-leading standards for low endotoxin content and superior results. Sodium bicarbonate is used as part of a buffering system commonly used to maintain physiological pH 7.2 - 7.4 in a culture environment. Bicarbonate is a weak buffer with that makes maintaining a pH range of 7.2 - 7.6 somewhat difficult, but it is non-toxic, and is a nutrient source. Sodium Bicarbonate #25-035 is offered in a 7.5% solution prepared in Water for Injection (WFI) quality water. See the addition Chart included in the Technical Information section to the left for information on proper use. - Complete Certificate of Analysis available for each lot, along with full formulation information
Corning media certificates are available on our Resource Library.
Corning media certificates are available on our Resource Library.
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Form | Liquid |
| Product Type | Sodium Bicarbonate |
| Sterility | Sterile-filtered |
| Format | Plastic Bottle |
| pH | 7.8 to 8.2 |
| Quantity | 6 x 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?