missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Penicillin-Streptomycin-L-Glutamine
BIOPHARMA

Product Code. 15373671
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 15373671

Brand: Corning™ 30009CI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Packaged in plastic bottles

Technical Advantage:  Effective against both Gram-positive and -negative bacteria
Penicillin-Streptomycin-L-glutamine combines two antibiotics with a nutrient which is essential for cell growth, L-glutamine.  Penicillin inhibits bacterial growth by inhibiting peptidoglycan synthesis (essential for the bacterial cell wall), and streptomycin inhibits bacterial growth by inhibiting protein synthesis.  L-Glutamine is essential as an energy source for most mammalian cells in culture because of its amino acid carbon skeleton.  This antibiotic-nutrient combination is effective against Gram-negative and Gram-positive bacteria.
-  Complete Certificate of Analysis available for each production lot, along with full formulation information
-  Produced under the highest industry standards to ensure superior results
10,000 I.U. Penicillin (per mL) 10,000 μg/ mL Streptomycin
Corning media certificates are available on our Resource Library.

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Concentration 10,000 I.U. Penicillin (per mL); 10,000 ug/mL Streptomycin; 29.2 mg/mL L-glutamine
Form Liquid
Product Type Antibiotic-L-Glutamine
Format Plastic Bottle
Quantity 6 x 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.