missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ Iscove's Modification of DMEM, Powder

Product Code. 15303741
Change view
Click to view available options
Quantity:
1 x 10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15303741 1 x 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15303741 Supplier Corning™ Supplier No. 50016PB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ cellgro™ Iscove's Modification of DMEM, Powder is hybridoma tested.

  • Highly enriched synthetic medium optimized
  • Rapidly proliferates cultures of high cell density
  • IMDM contains additional amino acids, vitamins, selenium and HEPES buffer
  • Formulated with potassium nitrate in place of ferric nitrate
  • Addition of albumin and transferrin may be required for certain cell types
  • Formulation contains 25mM HEPES
  • Does not contain alpha-thioglycerol, beta-mercaptoethanol and sodium bicarbonate
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Powder
Product Type IMDM
With Additives HEPES
Without Additives No α-Thioglycerol, No beta-Mercaptoethanol, No Sodium Bicarbonate
Quantity 1 x 10 L
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.