missing translation for 'onlineSavingsMsg'
Learn More

Corning™ HEPES, Liquid
BIOPHARMA

Product Code. 15323661
Change view
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15323661 6 x 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15323661 Supplier Corning™ Supplier No. 25060CI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An alternative to use of bicarbonate for buffering

Technical Advantage:  Product manufactured under industry-leading standards for low endotoxin content and superior results.
HEPES (N-2-hydroxyethylpiperazine-N'-2-ethanesulphonic acid) provides an alternative to the use of bicarbonate for buffering because it does not require elevated levels of CO2.  Unlike the low pKa of bicarbonate, HEPES has a pKa of 7.66 making it a much stronger buffer in the pH 7.2-7.6 range than bicarbonate buffer.  Potential disadvantages of HEPES include toxicity over 100 mM and lack of nutritive value to the cell.  The recommended working concentration is 10-25 mM.
-  Complete Certificate of Analysis available for each production lot, along with full formulation information

Corning media certificates are available on our Resource Library.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Form Liquid
Product Type HEPES Buffer
Sterility Sterile-filtered
pH 7.2 to 7.6
Quantity 6 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.