missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Gentamicin Sulfate
Packaged in plastic bottles
Brand: Corning™ 30-005-CR
Code : NEW
Additional Details : Weight : 0.10000kg
Specifications
15° to 30°C | |
Liquid | |
Sterile | |
10 x 10 mL |
50 mg/mL | |
Antibiotic | |
Plastic Bottle |