Learn More
Corning™ Flexible Packaging - Tubing Set
Product Code. 31932505 Shop All Corning Products
Change view
Click to view available options
Product Type:
Tubing Set
Unit Size:
Pack of 10
Product Code. Product Type Unit Size
31932505 Tubing Set Pack of 10
1 options
Product Code. 31932505 Supplier Corning™ Supplier No. 9170000
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Optional tubing sets are available in combination with all single-use bag options
36„ length 1/4„ I.D. clear chemically resistant, heat sealable, flexible tubing
Male Luer and female Luer cap
Female Luer and male Luer cap
Corning media certificates are available on our Resource Library.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tubing Set