Learn More
Corning™ Collection Bags
Product Code. 31932520 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
0.5 L
1 L
5 L
10 L
20 L
50 L
100 L
200 L
Film Description:
EVA
ULDPE
Unit Size:
Each
Pack of 15
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Film Description Unit Size
31932520 20 L EVA Pack of 15
17283574 1 L EVA Each
31932503 1 L EVA Pack of 15
17273574 0.5 L EVA Each
31932513 0.5 L EVA Pack of 15
16438983 5 L EVA Each
31932495 5 L EVA Pack of 15
16458983 20 L EVA Each
15796278 10 L EVA -
16448983 10 L EVA Each
31932502 10 L EVA Pack of 15
16468983 50 L EVA Each
31932518 50 L EVA Pack of 15
17213584 100 L ULDPE Each
17223584 200 L ULDPE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 31932520 Supplier Corning™ Supplier No. 9120047
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Collection bags with three ports used for biopharmaceutical applications
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
TRUSTED_SUSTAINABILITY
Specifications
Tubing Style Heat sealable flexible tubing
Film Description EVA
No. per Case 1/Ea.
No. per Pack 1/Pk.
Port Detail Port 1 - 0.37 in. Connector, 12 L x 0.37 I.D. x 0.62 in. O.D., chemically resistant, heat sealable flexible tubing, port 2 - male MPC connector with 0.37 in. barb, female MPC cap, port- 3 female MPC connector with 0.37 in. barb, male MPC plug, break-off i
Quantity Each
Capacity (Metric) 20 L