Learn More
Corning™ Endothelial Cell Growth Supplement (ECGS)
Ideal for endothelial cell culture
Brand: Corning™ 356006
Code : 67
Additional Details : CAS Number : 3810-74-0 Weight : 0.03000kg
Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
- Vialed, highly purified growth factors with high biological activity
- Broad range of highly purified natural and recombinant lymphokines
- Enriched culture supplements to be reconstituted/added in the medium of your choice
- Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
Specifications
2° to 8°C | |
50 to 150 µg/ mL depending on cell type. May require supplementation of 1 to 100 µg/mL heparin. | |
Lyophilized from NaCl-containing buffer with streptomycin sulfate; reconstitute in serum-free medium | |
Bovine brain |
ECGS is a broadly used supplement to culture a variety of cells, particularly endothelial cells. ECGS contains various growth factors (e.g., acidic FGF or ECGF-a) | |
Growth Factor | |
100 mg | |
Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma. |