Learn More
Corning™ Bovine Pituitary Extract (BPE)
Supplement used in the culture of a variety of epithelial and endothelial cells
Brand: Corning™ 356123
Code : 67
Additional Details : Weight : 0.02400kg
Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
- Vialed, highly purified growth factors with high biological activity
- Broad range of highly purified natural and recombinant lymphokines
- Enriched culture supplements to be reconstituted/added in the medium of your choice
- Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
Specifications
2° to 8°C | |
Bovine Pituitary Extract | |
75 mg |