Learn More
Corning™ Black 384-Well Polypropylene Assay Plate
Lower compound sticking and higher solvent resistance than polystyrene plates
Brand: Corning™ 3683BC
Code : A0
Additional Details : Weight : 5.26167kg
Description
- Ideal for fluorescence assays and for compound storage
- Round-bottom wells
- Untreated
Specifications
Standard | |
16 x 24 | |
Polypropylene | |
Sterile | |
Flat Bottom (F-well) |
With Lid | |
Black | |
384 | |
CellBIND | |
For fluorescence assays and compound storage |