Learn More
Corning™ BioCoat™ TC-treated Multiwell Variety Packs
Product Code. 10335631
Change view
Click to view available options
Item Description:
Variety pack I
Variety pack II
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Item Description unitSize
10335631 Variety pack II Pack of 5
10586571 Variety pack I Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10335631 Supplier Corning™ Supplier No. 354431
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Choose from different groups of cellware conveniently packaged together in popular combinations
  • Selections include different combinations of extracellular matrix proteins and attachment factors
Multiwell Plate Variety Packs
  • 6-well
  • Pack I contains five plates (clear, round well, flat-bottom, TC-treated, with 9.6cm2 cell growth area)
  • Coated with one of the following: Collagen I, Collagen IV, Fibronectin, Laminin, and Poly-D-Lysine
  • Lids (Fisher P/N 08-774-133 only)
TRUSTED_SUSTAINABILITY
Specifications
Type Cellware Variety Pack
Item Description Variety pack II
Storage Requirements 2° to 8°C