missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Product Code. 10262323
Change view
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10262323 Black, Clear Bottom Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10262323 Supplier Corning™ Supplier No. 356396

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid

 

  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Available in 6, 12, 24, 48, 96 and 384 wells

 

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Format Standard
Lid With Lid
Content And Storage 4°C to 30°C
Array 16 x 24
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
Quantity 50/Cs.
Source Poly-D-Lysine
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.