Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
GREENER_CHOICE
Product Code. 10399320
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Quantity:
10/cs.
100/cs.
20/cs.
40/cs.
5/cs.
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 40
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Quantity unitSize
10399320 35 mm 20/cs. Pack of 20
10478822 35 mm 100/cs. Pack of 100
10003860 60 mm 20/cs. Pack of 20
10145290 100 mm 10/cs. Pack of 10
10204281 60 mm 100/cs. Pack of 100
10632122 100 mm 40/cs. Pack of 40
10307432 150 mm 5/cs. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10399320 Supplier Corning™ Supplier No. 354467
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Coated with 0.2 micron filtered PDL in a cleanroom
Note: Dishes are not terminally sterilized and not assigned an SAL level. 
Order Info
Bulk quantities available; please contact your Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Round
Color Clear
Culture Area 9.6 sq. cm
Material Polystyrene
Shape Round
Surface Treatment Poly-D-Lysine
Type Cell Culture Dish
Includes Lid
Quantity 20/cs.
Diameter (Metric) 35 mm