missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Matrigel Matrix Multiwell Plates
GREENER_CHOICE

Product Code. 10346011
Change view
Click to view available options
Well Count:
6
24
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
10346011 24 Pack of 2
10717341 6 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346011 Supplier Corning™ Supplier No. 354433

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Lid

Cell culture microplate

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Culture Area 3.8 cm2
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment ECM/Attachment Factors
Type Multi-well Cell Culture Plate
Well Count 24
For Use With (Application) Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Includes Lid
No. per Case 2/Cs.
No. per Pack 1/Pk.
Storage Requirements -20°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.