missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates
GREENER_CHOICE

Product Code. 10681274
Click to view available options
Well Count:
6
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 10681274

Brand: Corning™ 354671

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Each lot of plates is tested for the amount of functional proteins coated onto the 6-well plates to ensure lot-to-lot consistency.

Quality

  • Each lot of plates is tested for the amount of functional proteins coated onto the 6-well plates to ensure lot-to-lot consistency.
  • Functional performance was validated by culturing undifferentiated human embryonic stem cells (H9, H14, or H1 from WiCell Institute) in combination with mouse embryonic fibroblast (MEF) conditioned media (CM) supplemented with 8 ng/ml of basic FGF.
  • Tested and found negative for bacteria and fungi.

Features

  • Improved Transfection Efficiencies of hES Cells
  • Easier DNA, RNA, and Protein Isolation
  • Quality Assurance Tested
  • Ready-to-Use Convenience
  • Optimized culture protocol
  • Maintain Only One Cell Type

Preparation and Storage
Stable for at least three months from date of shipment when stored at -20°C.

Application
When cultured on Corning Matrigel Matrix, hES cells:

  • Maintain normal karyotype
  • Demonstrate a stable proliferation rate and high telomerase activity
  • Express characteristic undifferentiated hES cell markers
  • Form teratomas in severe combined immunodeficient (SCID) mice and differentiate into cells from all three germ layers

For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.

TRUSTED_SUSTAINABILITY

Specifications

Array 2 x 3
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Cell Culture
Type Multi-well Cell Culture Plate
Well Count 6
For Use With (Application) ES Culture
Includes Lid
No. per Case 5/Cs.
No. per Pack 5/Pk.
Quantity 5/Cs.
Source EHS Mouse Tumor
Storage Requirements -20°C
Show More Show Less

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.