missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates

Product Code. 10142561
Change view
Click to view available options
Culture Area:
0.75 sq. cm
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area unitSize
10142561 0.75 sq. cm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10142561 Supplier Corning™ Supplier No. 354508

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Corning Matrigel Basement Membrane Matrix is a solubilized tissue basement membrane preparation containing laminin, collagen type IV, heparan sulfate proteoglycan and entactin. Corning Matrigel Matrix is useful as a substrate for growth and differentiation of many normal and transformed cell types, such as epithelial, endothelial, muscle and neuronal cells

Corning BioCoat Thin Layer Matrigel cultureware contain a dried thin layer of Corning Matrigel Basement Membrane Matrix. Rehydration results in the formation of a thin “basement membrane” gel layer on the cultureware surface. Specific applications for Thin Layer Matrigel cellware include culturing of primary contractile skeletal myotubes, neuronal cell morphological differentiation, studies of vascular smooth muscle cell interaction with the extracellular matrix and the effects of the extracellular matrix on the function and morphology of lung alveolar, and bile ductular epithelial cells.

  • Especially suited for culture of polarized cells, such as epithelial cells, and promotes differentiation of hepatocytes, mammary epithelial cells and smooth muscle cells
  • Promotes capillary tubule formation of endothelial cells and neurite regeneration
  • Can be used for angiogenesis assays
  • Tested for the ability to promote neurite outgrowth from chick dorsal root ganglia (in the absence of NGF)

TRUSTED_SUSTAINABILITY

Specifications

Array 6 x 8
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Culture Area 0.75 sq. cm
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment ECM/Attachment Factors
Type Multi-well Cell Culture Plate
Well Count 48
For Use With (Application) Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Includes Lid
Source EHS Mouse Tumor
Storage Requirements -20°C
Show More Show Less

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.