missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Laminin TC-treated Culture Dishes

Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells

Brand:  Corning™ 354553

 View more versions of this product

Product Code. 10604042

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.

Applications include:

  • Promotion of cell attachment and spreading
  • Culture of normal and transfected F9 teratocarcinoma cells for gene expression studies
  • Culture of HUVEC for E-Selectin7 expression and VEGF induction8

Source

  • Gelatin, porcine
  • Quality Control

    • Tested for ability to promote proliferation of HUVECs
    • Tested and found negative for bacteria and fungi

    Storage

  • 4°C to 30°C under dry conditions.
  • Order Info

    Bulk quantities available; please contact your Customer Service Representative.

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Round
    Polystyrene
    Cell Culture Dish
    150 mm
    Clear
    Round
    Laminin
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers