missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Laminin TC-treated Culture Dishes

Product Code. 10192651
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10192651 35 mm Pack of 20
10448822 60 mm Pack of 20
10526381 100 mm Pack of 10
10604042 150 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10192651 Supplier Corning™ Supplier No. 354458

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells

Laminin (LM), a major component of basement membranes, is a multifunctional glycoprotein that is used as a substrate to culture and maintain differentiated function of a wide variety of cells. Laminin has been shown in culture to stimulate neurite outgrowth, promote cell attachment, chemotaxis, cell differentiation, and neuronal survival.

Applications include:

  • Promotion of cell attachment and spreading
  • Induction of cell differentiation and neurite outgrowth
  • Increases proliferation of myoblasts
  • Studies of effects of laminin on cell behavior
  • Cell adhesion assays

Source

  • Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Quality Control

    • Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells
    • Tested and found negative for bacteria and fungi
    • Laminin purity >90% by SDS-PAGE (contains entactin)

    Storage

  • 2°C to 8°C. Do not freeze.
  • Order Info

    Bulk quantities available; please contact your Customer Service Representative.

    TRUSTED_SUSTAINABILITY

    Specifications

    Bottom Shape Round
    Color Clear
    Culture Area 9.6 cm2
    Material Polystyrene
    Shape Round
    Type Cell Culture Dish
    No. per Case 20/Cs.
    No. per Pack 10/Pk.
    Supportive Membrane Laminin
    Diameter (Metric) 35 mm
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.