missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Gelatin Multiwell Plate
GREENER_CHOICEBIOPHARMA

Product Code. 10237430
Change view
Click to view available options
Culture Area:
9.6 cm2
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area unitSize
10237430 9.6 cm2 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10237430 Supplier Corning™ Supplier No. 354652

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Promotion of cell attachment and spreading

Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.

Applications include:

  • Promotion of cell attachment and spreading
  • Culture of normal and transfected F9 teratocarcinoma cells for gene expression studies
  • Culture of HUVEC for E-Selectin7 expression and VEGF induction8

Source

  • Gelatin, porcine
  • Quality Control

    • Tested for ability to promote proliferation of HUVECs
    • Tested and found negative for bacteria and fungi

    Storage

  • 4°C to 30°C under dry conditions.
  • Order Info

    Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

    TRUSTED_SUSTAINABILITY

    Specifications

    Array 2 x 3
    Closure Color Clear
    Closure Included Yes
    Closure Material Polystyrene
    Bottom Shape Flat
    Color Clear
    Culture Area 9.6 cm2
    Material Polystyrene
    Shape Rectangular
    Sterility Non-sterile
    Surface Treatment Cell Culture
    Type Multi-well Cell Culture Plate
    Well Count 6
    For Use With (Application) Promote cell attachment, spreading, proliferation and differentiation
    Includes Lid
    No. per Case 5/Cs.
    No. per Pack 5/Pk.
    Quantity 5/Cs.
    Source Gelatin
    Storage Requirements 4°C to 30°C
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.