missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Fibronectin TC-treated Culture Dish
GREENER_CHOICE

Product Code. 10747351
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10747351 100 mm Pack of 10
10214091 35 mm Pack of 20
10080741 60 mm Pack of 20
10152801 150 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10747351 Supplier Corning™ Supplier No. 354451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Matrix components are applied as an even, optically clear coating

  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody.

For adhesion, growth and differentiation of neuronal and transfected cell lines

Order Info

Bulk quantities available; please contact your Customer Service Representative.

TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Round
Color Clear
Culture Area 58.1 cm2
Material Polystyrene
Shape Round
Type Cell Culture Dish
No. per Case 10/Cs.
No. per Pack 5/Pk.
Diameter (Metric) 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.