missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Fibronectin-Treated, Flat-Bottom Microplate
GREENER_CHOICEBIOPHARMA

Product Code. 10738692
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10738692 Clear Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
An item in this group is an exact match for part number 0877460
Product Code. 10738692 Supplier Corning™ Supplier No. 354409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
An item in this group is an exact match for part number 0877460

Color:Clear; Sterility:Sterile; Lid:With Lid

Corning™ BioCoatTM Fibronectin 96-well Clear Plate is comprised of clear TC-treated polystyrene with a uniform application of human fibronectin. Flat bottom, with lid, 5/case

Corning BioCoat products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.

Order Info

Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Content And Storage 2°C to 8°C
Culture Area 0.32 cm2
Array 8 x 12
Packaging 5/Pk.
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Fibronectin
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays
Quantity 5/Cs.
Source Human Fibronectin
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.