missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Fibronectin Multiwell Plate

Product Code. 10040781
Change view
Click to view available options
Culture Area:
0.75 cm2
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area unitSize
10040781 0.75 cm2 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Product Code. 10040781 Supplier Corning™ Supplier No. 354506

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology Supplied with preapplied extracellular matrices, facilitating cell adaption to in vitro cultivation
  • Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo
  • Low pore-density, track-etched polyethylene teraphthalate (PET) membranes offer convenient viewing of living cells with a phase-contrast microscope
  • Have low-binding properties that minimize small molecule loss

The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody. These products are for research use only and are not intended for diagnostic purposes.

Order Info

Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

TRUSTED_SUSTAINABILITY

Specifications

Array 6 x 8
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Culture Area 0.75 cm2
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Tissue Culture
Type Multi-well Cell Culture Plate
Well Count 48
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays
Includes Lid
Source Human Fibronectin
Storage Requirements 2°C to 8°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.