missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis

Product Code. 10067911
Change view
Click to view available options
No. of Wells:
24
96
Culture Area:
0.08 cm2
0.3 cm2
Quantity:
1
5
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Culture Area Quantity unitSize
10067911 24 0.3 cm2 1 Each
10678892 24 0.3 cm2 5 Pack of 5
10396602 96 0.08 cm2 5 Pack of 5
10778124 96 0.08 cm2 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10067911 Supplier Corning™ Supplier No. 354143

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid

Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays

HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.

Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system

TRUSTED_SUSTAINABILITY

Specifications

Culture Area 0.3 cm2
Pore Size 3 μm
Type Multiwell Insert Plate
For Use With (Application) High Throughput Screening
Quantity 1
Surface Coating Fibronectin
No. of Wells 24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.