Learn More
Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis
Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays
Brand: Corning™ 354147
Code : 67
Additional Details : Weight : 0.15000kg
Includes: A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
0.08 cm2 | |
Multiwell Insert Plate | |
1 | |
96 |
3 μm | |
High Throughput Screening | |
Fibronectin |