Learn More
Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis

Description
Includes
A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
Specifications
| Culture Area | 0.3 cm2 |
| Material | PET |
| Pore Size | 3 μm |
| Type | Multiwell Insert Plate |
| For Use With (Application) | High Throughput Screening |
| Quantity | 5 |
| No. per Pack | 5/Pk. |
| No. per Case | 5/Cs. |
| Surface Coating | Fibronectin |
| No. of Wells | 24 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.