Learn More
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
GREENER_CHOICE
Product Code. 10185760
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Quantity:
10/cs.
100/cs.
20/cs.
40/cs.
5/cs.
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 40
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Quantity unitSize
10185760 150 mm 5/cs. Pack of 5
10555421 60 mm 100/cs. Pack of 100
10772531 35 mm 100/cs. Pack of 100
10155670 35 mm 20/cs. Pack of 20
10543051 60 mm 20/cs. Pack of 20
10572701 100 mm 10/cs. Pack of 10
10499012 100 mm 40/cs. Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10185760 Supplier Corning™ Supplier No. 354551
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Order Info
Bulk quantities available; please contact your Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Round
Color Clear
Culture Area 151.9 sq. cm
Material Polystyrene
Shape Round
Surface Treatment Collagen Type I
Type Cell Culture Dish
Includes Lid
Quantity 5/cs.
Diameter (Metric) 150 mm