missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 384-Well, Collagen Type I-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Product Code. 10393463
Change view
Click to view available options
Packaging:
80/Cs
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
10393463 80/Cs Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10393463 Supplier Corning™ Supplier No. 356705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Color: Black, Clear Bottom; Sterility: Non-sterile; Lid: With Lid

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SBS
    Format Standard
    Lid With Lid
    Content And Storage 4°C to 30°C
    Array 16 x 24
    Packaging 80/Cs
    Color Black, Clear Bottom
    Material Polystyrene
    No. of Wells 384
    Sterility Non-sterile
    Surface Treatment Collagen Type I
    Well Shape Round, Flat Bottom (F-well)
    For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
    No. per Case 80/Cs.
    No. per Pack 20/Pk.
    Quantity 80/Cs.
    Source Rat Tail
    Well Color Clear
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.