missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ 384-Well, Collagen Type I-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile
Brand: Corning™ 354397
This item is not returnable.
View return policy
Description
- Source: Rat tail
- TC-treated
- Clear, round well
- Flat bottom
- Nonsterile
For research use only.
Specifications
ANSI, SBS | |
4°C to 30°C | |
5/Pk., 50/Cs. | |
Low Volume | |
384 | |
Collagen Type I | |
Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture | |
Rat Tail |
Standard | |
16 x 24 | |
Black, Clear Bottom | |
Polystyrene | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
50/Cs. | |
Clear |