missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
Description
- Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Order Info
Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
| Pore Size | 0.3 μm |
| Type | Cell Culture Insert |
| For Use With (Application) | Adhesion, growth and differentiation of neuronal and transfected cell lines |
| Quantity | 1 |
| Storage Requirements | 2° to 8°C |
| No. of Wells | 24 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?