Learn More
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand: Corning™ 354482
Code : 67
Additional Details : Weight : 0.23000kg
Description
- Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Order Info
Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.
Specifications
12 | |
1 μm | |
Adhesion, growth and differentiation of neuronal and transfected cell lines | |
2° to 8°C |
PET | |
Well Plate | |
2 |