Learn More
Corning™ BioCoat™ Assay System
Product Code. 11513590
Change view
Click to view available options
Type:
Cell Culture Insert System
Multi-well Insert System
Quantity:
1/Cs.
5/Cs.
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity unitSize
11513590 Cell Culture Insert System 1/Cs. Each
10631932 Cell Culture Insert System 5/Cs. Pack of 5
10596951 Multi-well Insert System 5/Cs. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11513590 Supplier Corning™ Supplier No. 354801
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
Automation-friendly design
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Culture Area 0.3 cm2
Sterility Non-sterile
Surface Treatment Collagen
Type Cell Culture Insert System
Well Count 24
For Use With (Application) Rapid differentiation of Caco 2 cells in vitro for robotic screening of prospective pharmaceuticals for oral bioavailability and absorption
Includes 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid
Pore Size 1 μm
Quantity 1/Cs.