Learn More
Corning™ BioCoat™ Assay System
Automation-friendly design
Brand: Corning™ 354804
Code : 66
Additional Details : Weight : 0.40000kg
Includes: 24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
Yes | |
0.3 cm2 | |
Non-sterile | |
Multi-well Insert System | |
Falcon™ companion plate | |
5/Ea. | |
1 μm | |
Polyethylene Terephthalate |
Clear | |
PET | |
Collagen | |
24 | |
24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid | |
1/Pk. | |
5/Cs. |