Learn More
Corning™ Adapters for LSE™ Compact Centrifuges
For use with Corning LSE high speed microcentrifuge.
Brand: Corning™ 480135
Code : NEW
Additional Details : Weight : 0.05000kg
Description
The individual adaptors for tubes are used in the Corning LSE High Speed Microcentrifuge.
Specifications
Adapter | |
0.2 mL tubes | |
6/Pk. |