Learn More
Corning™ Adapters for LSE™ Compact Centrifuges
For use with Corning LSE high speed microcentrifuge.
Brand:  Corning™ 480140
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 15703338
  • £113.38 / Pack of 6
Estimated Shipment: 18-12-2024
to see stock.
Description
Description
The individual adaptors for tubes are used in the Corning LSE High Speed Microcentrifuge.
Specifications
Specifications
Adapter
10/15 mL tubes in 50 mL rotor
6/Pk.