missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Product Code. 10443192 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10443192 Black, Clear Bottom Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10443192 Supplier Corning™ Supplier No. 3720

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Color:Black, Clear Bottom; Sterility:Sterile; Lid:Without Lid

  • Clear well bottoms are 60% thinner than conventional plates for direct viewing from either top or bottom
  • Black opaque walls emit minimal background for sharp, clear images of fluorescent cells
  • Available untreated or with tissue culture-treated surfaces

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Spectroscopy, polarization and imaging
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.