missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Product Code. 10441052 Shop All Corning Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10441052 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10441052 Supplier Corning™ Supplier No. 3362
  • ONLINE EXCLUSIVE PRICE
    £595.00
    £555.98 / Pack of 100
    Save £39.02
    6% Off
Estimated Shipment: 23-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color:White; Sterility:Sterile; Lid:Without Lid

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Lid Without Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Color White
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Treated for optimal cell attachment
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.