missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Product Code. 10367382 Shop All Corning Products
Change view
Click to view available options
Color:
White, Clear Bottom
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10367382 White, Clear Bottom Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10367382 Supplier Corning™ Supplier No. 3885
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:White, Clear Bottom; Sterility:Sterile; Lid:With Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Half-Area
    Lid With Lid
    Culture Area 0.16 cm2
    Array 8 x 12
    Color White, Clear Bottom
    For Use With (Equipment) Standard automated handling and detection systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Homogenous and cell based assays
    No. per Case 100/Cs.
    No. per Pack 20/Pk.
    Quantity 100/Cs.
    Well Color Clear
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.