missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molded polystyrene with opaque walls and optically clear bottoms
- Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
- Opaque black or white walls prevent well to well crosstalk
- Recommended working volumes of 20 to 80μL; total well volume of 112μL
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Suited for flourescent and luminescent assays
- Plate dimensions meet proposed industry standard
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.06 cm2 |
| Array | 16 x 24 |
| Color | White, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Sterility | Sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?