missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate

Product Code. 10193661 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10193661 Black, Clear Bottom Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10193661 Supplier Corning™ Supplier No. 3712

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

 

  • Molded polystyrene with opaque walls and optically clear bottoms
  • Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
  • Opaque black or white walls prevent well to well crosstalk
  • Recommended working volumes of 20 to 80μL; total well volume of 112μL
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Suited for flourescent and luminescent assays
  • Plate dimensions meet proposed industry standard
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid With Lid
Culture Area 0.06 cm2
Array 16 x 24
Packaging 20/Pk.
Color Black, Clear Bottom
Material Polystyrene
Volume (Metric) Well 112 μL
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 112 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
Quantity 100/Cs.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.