missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Compatible with high throughput confocal imagers
- Better cell growth and attachment than glass bottom plates
- TC-treated for optimal cell attachment
Generic Barcode
- Ensures compatibility with most bar code scanning and software systems
- To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
- Labels applied to all 4 sides of the plate
- No sequence number duplicates
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.05 cm2 |
| Array | 16 x 24 |
| Packaging | 20/Pk., 100/Cs. |
| Color | Black, Clear Bottom |
| Product Type | Optical Imaging |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?