missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
- Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
- Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
- Choose black plates for fluorescence applications or white for luminescence applications
- Ideal for immunoassays
- Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
- Working volume: 2 to 20μL
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Array | 16 x 24 |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 5 to 40 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-binding |
| Well Shape | Round |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?