missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE

Product Code. 10359854 Shop All Corning Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10359854 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10359854 Supplier Corning™ Supplier No. 3576

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.

Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Binding Type High Binding
Color White
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
For Use With (Application) Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.