Learn More
Corning™ 384-Well Assay Multiwell Plates
Shop All Corning Products
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 100
Description
Sold in bulk pack
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Color | Black, Clear Bottom |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Surface Treatment | Collagen |