missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Assay Multiwell Plates
Shop All Corning Products
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 100
Description
Sold in bulk pack
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Packaging | 20/Pk., 100/Cs. |
| Color | Black, Clear Bottom |
| Product Type | 384-Well Assay Multiwell Plates |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | Fibronectin |
| Well Shape | Flat Bottom (F-well) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction